missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Disodium Salt, Dihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
Supplier: Avantor J.T.Baker 404001
Description
- For Molecular Biology Applications
Specifications
| EDTA, Disodium Salt | |
| 6381-92-6 | |
| C10H18N2Na2O10 | |
| 500 g | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| Ultrapure Bioreagent |
| Dihydrate | |
| Poly Bottle | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| 372.24 | |
| Crystals |
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |