missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Disodium Salt, Dihydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02002789
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002789 100 g Poly Bottle
02002787 250 g Poly Bottle
02002790 500 g Poly Bottle
02002791 1 kg Poly Bottle
02002788 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02002789 Supplier Avantor Supplier No. 404000

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Packaging Poly Bottle
Quantity 100 g
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24
Grade Ultrapure Bioreagent
Physical Form Crystals
Chemical Name or Material EDTA, Disodium Salt, Dihydrate