missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5383269
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002786 500 g Poly Bottle
02002785 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002786 Supplier Avantor Supplier No. 899301
Only null left

High quality chemicals for laboratory and specialized industrial use

    Bulk Pharmaceutical Chemical

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 0.00kg
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24
Grade Meets USP requirements
Physical Form Crystals
Chemical Name or Material EDTA Disodium Salt, Dihydrate