missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt concentrate, For 1L standard solution, 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™

for 1L standard solution, 0.1 M EDTA-Na2, for complexometry

$136.71 - $715.24

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002662
View Documents
Honeywell Chemicals
380571EA
1 Pc.
Each for $136.71
Only null left
 
60046683
View Documents
Honeywell Chemicals
380576X1EA
6 x 1 Ea.
Pack of 6 Each for $715.24
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

139-33-3
336.21
ZGTMUACCHSMWAC-UHFFFAOYSA-L
CHEBI:64734
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H14N2Na2O8
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
57339238
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate

Specifications

139-33-3
C10H14N2Na2O8
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
3822669
ZGTMUACCHSMWAC-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
57339238
336.21g/mol
Ethylenediaminetetraacetic acid disodium salt concentrate
1 Pc.
[-CH2N(CH2CO2Na)CH2CO2H]2
UN3265
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
336.21
CHEBI:64734
Ampoule
Videos
Safety and Handling

Safety and Handling

P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

ShelfLife : 1260 days from date of manufacture

DOTInformation : 8 / PGII

Recommended Storage : Room Temp

SDS
Documents

Documents