missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents

Catalog No. 01337261
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337261 10 kg
01337259 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337261 Supplier Labchem Inc Supplier No. C220110010KG
Only null left

EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material EDTA Disodium Dihydrate, Purified ((Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate)
CAS 6381-92-6
Boiling Range >252°C
Melting Point 252°C
Color White
pH 4 to 6
Boiling Point >252°C
Melting Range 252°C
Packaging Poly Pail
Molecular Formula C10H14N2O8Na2, 2H2O
Quantity 10 kg
Specific Gravity 1
Synonym Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate
Molecular Weight (g/mol) 372.2369
Grade Pure
Physical Form Solid
Show More Show Less