missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Disodium, Dihydrate, ACS Grade, 99.0 to 101.0%, LabChem™
Specifications
Specifications
| CAS Max % | 100 |
| Melting Point | 252°C |
| Color | White |
| Packaging | Poly Bottle |
| Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O |
| Quantity | 100 g |
| Solubility Information | Soluble in water |
| Formula Weight | 372.23 |
| Percent Purity | 99.0 to 101.0% |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. Keep container tightly closed. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If on skin: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up in a well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| EINECSNumber | 205-358-3 |
| Recommended Storage | Room Temperature |