missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA dipotassium magnesium salt, For complexometry, ≥98%, Honeywell Fluka™
for complexometry,≥98%
$263.58 - $1376.53
Chemical Identifiers
| CAS | 15708-48-2 |
|---|---|
| Molecular Formula | C10H12K2MgN2O8 |
| Molecular Weight (g/mol) | 390.714 |
| MDL Number | MFCD00054450 |
| InChI Key | MUEOBEUHFKBRJH-UHFFFAOYSA-J |
| Synonym | dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2- |
| PubChem CID | 85061 |
| IUPAC Name | magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate |
| SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 15708-48-2 | |
| 390.714 | |
| MUEOBEUHFKBRJH-UHFFFAOYSA-J | |
| 85061 | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+] |
| C10H12K2MgN2O8 | |
| MFCD00054450 | |
| dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2- | |
| magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate |
Specifications
| 15708-48-2 | |
| C10H12K2MgN2O8 | |
| NONH for all modes of transport | |
| MUEOBEUHFKBRJH-UHFFFAOYSA-J | |
| magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate | |
| 85061 | |
| ≥98% |
| 100 g | |
| MFCD00054450 | |
| dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2- | |
| C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+] | |
| 390.714 | |
| 426.74g/mol | |
| Ethylenediaminetetraacetic acid dipotassium magnesium salt |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
EINECSNumber : 239-803-8
Recommended Storage : Room Temp