missing translation for 'onlineSavingsMsg'
Learn More

EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents

Catalog No. p-200021068
Change view
Click to view available options
Quantity:
125 g
500 g
10 lb.
10 kg
50 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337988 10 lb.
01337987 10 kg
01337986 125 g
01337985 500 g
01337984 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337988 Supplier Labchem Inc Supplier No. C120110010F5
Only null left

EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym Edetate Disodium, Disodium EDTA, (Ethylenedinitrilo)tetraacetic Acid disodium salt dihydrate
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 252°C
Color White
pH 4 to 6
Boiling Point >252°C
Packaging Poly Pail
Quantity 10 lb.
Specific Gravity 0.86
Grade ACS
Physical Form Solid
Chemical Name or Material EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate)