missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA (Dihydrate), ACS Reagent (Ethylenedinitrilo)tetraacetic Acid, Disodium Salt Dihydrate), Reagents
Specifications
Specifications
| Melting Point | 252°C |
| Color | White |
| pH | 4 to 6 |
| Boiling Point | >252°C |
| Packaging | Poly Pail |
| Quantity | 10 lb. |
| Specific Gravity | 0.86 |
| Percent Purity | ≥99% |
| Grade | ACS |
| Physical Form | Solid |
| Show More |