missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Certified, 0.1000M ±0.0005M (0.2N), LabChem™
Supplier: LabChem LC139004
Specifications
| EDTA, 0.1M (0.2N) | |
| 96.28,3.72 | |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 44120005 | |
| Certified | |
| Liquid |
| 6381-92-6,7732-18-5 | |
| Poly Bottle | |
| (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O | |
| 4 L | |
| Soluble in water | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| 372.23 | |
| 0.1000M ±0.0005M (0.2N) |
Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| 44120005 | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature