missing translation for 'onlineSavingsMsg'
Learn More

EDTA, Certified, 0.0050M ±0.0001M (0.01N), LabChem™

Supplier:  LabChem LC138202

Catalog No. LC138202

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

EDTA
99.81,0.19
1g/mL
C10H18N2Na2O10
MFCD00150037,MFCD00003541
1g/mL
Soluble in water
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.23
0.0050M ±0.0001M (0.01N)
6381-92-6,7732-18-5
Colorless
Poly Bottle
(HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
1 L
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
44120005
Certified
Liquid

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions