missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Edge Utility Cutter

Catalog No. 17010318
Encompass
Change view
Click to view available options
Color:
Black, Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17010318 Black, Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17010318 Supplier Slice™ Supplier No. 10479

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Edge Utility Cutter is well crafted and sturdy. Ideal for any cut that requires a clamping motion, due to its smooth and steady movement. The comfortable soft-touch handle includes a Blade Garage™ for extra capacity and a lock for safe tool storage.

With contoured, soft-touch grips and a sturdy metal handle, the Edge Utility Cutter fits in your hand perfectly and is easy to use. When you want to change blades, simply utilize the tool's clamp-tightening screw to switch them out – no tools required. Additionally, you can store multiple blades in the handle's Blade Garage™, ensuring your workflow is never interrupted.

From cutting heavy-duty zip ties to chopping through wire and rubber, Slice makes people and places safer. Learn more below.

  • Durable steel handle with soft-touch comfort grips
  • Blade guard and tool lock provide safe storage
  • 100% zirconium oxide blade that never rusts
  • Blade lasts on average 11.2 times longer than metal blades
  • Easy-to-use ambidextrous design

Great for:

  • Cutting wire and rubber tubing
  • Cutting heavy-duty zip ties
  • Cutting thin extruded plastics

Specifications

Blade Material Zirconium Oxide
Product Type Cutter
For Use With (Application) Cutting wire and rubber tubing, heavy-duty zip ties, thin extruded plastics
Length (English) Blade 0.28 in.
Length (Metric) Blade 7 mm
Size 9.65 x 2.5 x 1.08 in.
Sterility Non-sterile
Includes Slice Blade 10524
Handle Material TPR, GFN, POM, Carbon Steel, Stainless Steel
Color Black, Orange
Packaging Type Corrugated box
Show More Show Less