missing translation for 'onlineSavingsMsg'
Learn More

Edge Biologicals Nutrient Agar Slant

Catalog No. EBT0466
Encompass
Change view
Click to view available options
Form:
Slant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
EBT0466 Slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EBT0466 Supplier Edge Biologicals Supplier No. T0466
Only null left

Sterile and autoclavable general culture medium

Clear amber suspension is useful for the cultivation of the majority of less fastidious microorganisms.

Composition (g/L):

  • Beef Extract, 3g
  • Peptone, 5g
  • Sodium chloride, 8g
  • Agar, 15g
  • Deionized water, 1000 mL

Laboratory Use Only. Not for use in diagnostic or therapeutic procedures

Specifications

Form Slant
Manufacturing Origin United States
Media Type Nutrient Agar
Product Type Agar
For Use With (Application) Cultivation of many of the less fastidious microorganisms
Packaging Type Tube
Quantity 10/Pk.