missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-929 Double Layer Nitrile Coated Gloves

Catalog No. 19501073
Change view
Click to view available options
Size:
10
11
12
6
7
8
9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19501073 12
19501067 6
19501068 7
19501069 8
19501070 9
19501071 10
19501072 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19501073 Supplier Ansell Supplier No. 48929120

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Double layer NBR coating keeps hands dry and comfortable, in addition to enhancing grip capabilities when handling oily parts and tools.

  • Fully dipped NBR coating keeps hands dry and comfortable when handling oily parts and tools
  • Textured palm enhances grip capabilities
  • Seamless liner is comfortable and flexible
  • Elastic knitwrist ensures a secure fit

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Seamless Knit
Coating Material Nitrile
Color Black, Gray
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Fiberglass, Polyethylene, Spandex
Product Type Dip-coated Gloves
Size 12
EN388 Standard 4X43C
For Use With (Application) Aerospace, Automotive Assembly, Chemicals, Glass Manufacturing, Metal Fabrication, and Transportation
Glove Cuff Style Knit
Series 48-929
Yarn Gauge 13
Show More Show Less