missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-929 Double Layer Nitrile Coated Gloves

Catalog No. 19501069
Change view
Click to view available options
Size:
10
11
12
6
7
8
9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19501069 8
19501067 6
19501068 7
19501070 9
19501071 10
19501072 11
19501073 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19501069 Supplier Ansell Supplier No. 48929080
Only null left

Double layer NBR coating keeps hands dry and comfortable, in addition to enhancing grip capabilities when handling oily parts and tools.

  • Fully dipped NBR coating keeps hands dry and comfortable when handling oily parts and tools
  • Textured palm enhances grip capabilities
  • Seamless liner is comfortable and flexible
  • Elastic knitwrist ensures a secure fit

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Seamless Knit
Coating Material Nitrile
Color Black, Gray
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Fiberglass, Polyethylene, Spandex
Product Type Dip-coated Gloves
Size 8
EN388 Standard 4X43C
For Use With (Application) Aerospace, Automotive Assembly, Chemicals, Glass Manufacturing, Metal Fabrication, and Transportation
Glove Cuff Style Knit
Series 48-929
Yarn Gauge 13
Show More Show Less