missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetic Acid, NF, 98-100.5%, Spectrum™ Chemical
E1208, 60-00-4, C10H16N2O8
Supplier: Spectrum Chemical Mfg Cor E120825KGBL
Description
Spectrum™ Chemical Edetic Acid, NF is used as a chelating agent in therapy and as preservative adjuncts in pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 100% | |
| Fiber Drum | |
| MFCD00003541 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid | |
| 98 to 100.5% |
| 0.2% | |
| C10H16N2O8 | |
| 25 kg | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
| 292.24 | |
| NF |
Chemical Identifiers
| C10H16N2O8 | |
| MFCD00003541 | |
| 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid |
| 292.24 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |