missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetate Disodium Dihydrogen, Dihydrate, Primary Standard, 99.9-100.1%, Spectrum™ Chemical
E1247, 6381-92-6, C10H14N2Na2O8·2H2O
$538.94 - $1308.66
Chemical Identifiers
| CAS | 6381-92-6 |
|---|---|
| Molecular Formula | C10H18N2Na2O10 |
| Molecular Weight (g/mol) | 372.24 |
| MDL Number | MFCD00150037,MFCD00003541 |
| InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
| SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Description
Spectrum™ Chemical Edetate Disodium Dihydrogen, Dihydrate, Primary Standard is a single component organic chemical reference standard. Reference standards are particularly important for analytical chemistry since most analytical instrumentation is comparative; it requires a sample of known composition (reference standard) for accurate calibration. These reference standards are produced under stringent manufacturing procedures and differ from laboratory reagents in their certification and the traceability of the data provided.Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Specifications
| 6381-92-6 | |
| Poly Bottle | |
| MFCD00150037,MFCD00003541 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 99.9 to 100.1% |
| 100% | |
| C10H18N2Na2O10 | |
| 25 g | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| Primary Standard |