missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edetate Disodium, Dihydrate, USP, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Edetate Disodium, Dihydrate, USP is a chelating agent used to control heart rhythm disturbances caused by a heart medications. All Spectrum Chemical USP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Loss on Drying | 8.7-11.4% |
| Packaging | Poly Bottle |
| Quantity | 125 g |
| Percent Purity | 99 to 101% |
| Grade | USP |