missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Eddy Current Unit

Catalog No. S12980
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S12980 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12980 Supplier Eisco™ Supplier No. PH0845

Apparatus to demonstrate Lenz law.

  • Consists steel mass and neodymium magnet mass dropping down 33cm copper tube
  • Eddy current is explained when students see magnetic mass fall more slowly than steel mass
  • Eddy currents oppose downward motion through moving magnetic field created by falling neodymium magnet
  • Includes test masses

Specifications

For Use With (Application) Demonstrates Lenz law
Grade 9 to 12
Product Type Eddy Current Unit
Includes Steel and neodymium magnet masses