missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Eddy Current Demonstrator

Catalog No. S07376
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07376 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07376 Supplier United Scientific™ Supplier No. EDYCR1

Use this Eddy Current Demonstrator, a simple but effective device, to demonstrate Lenz's Law.

  • Uses eddy currents induced in the wall of a copper tube by a falling magnet.
  • Consists of 12 in. (30.4 cm) long copper tube and two apparently identical metal slugs (steel and neodymium magnet) with rubber end caps.
  • Plain steel slug falls through copper tube quickly but neodymium magnet falls very slowly due to braking effect of eddy currents.
  • Includes tube end caps for storing slugs in tube and an activity guide.

Specifications

Material Metal
Shape Cylindrical
For Use With (Application) Demonstrate Lenz's law with this simple but effective device
Grade 5+
Product Type Eddy Current Demonstrator
Includes Tube end caps for storing the slugs in the tube and an activity guide