missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDAC, 1-Ethyl-3-(3-Dimethylaminopropyl)carbodiimide, Hydrochloride
Description
Protocols for crosslinking molecules with this reagent can be found in Bioconjugate Techniques, by G.T. Hermanson (B-7884).
Specifications
Specifications
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 191.7 |
| Physical Form | Solid |
Frequently Asked Questions (FAQs)
Is your EDAC compound the same as your EDC compound?
Yes, both acronyms are used for the same compound. The chemical name for EDC is 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide and the chemical name for EDAC is N-3-(3-dimethylaminopropyl)-N-ethyl-carbodiimide. Both of these compounds are structurally equivalent.
For Research Use Only. Not for use in diagnostic procedures.