EcoSeal™ features smaller dimensions and a single non-perforated end-tab to provide a more cost-effective alternative to protect samples from evaporation, external contamination, dust and particulates. Provides a secure seal and is easy to remove. Rigid polyester film features good clarity and easy handling. Can be used as a temporary seal while awaiting further processing.