Learn More
LGC Genomics Llc (Lucigen) EconoTaq DNA Polymerase (separate Mg++), 1,000 UDependable Taq polymerase performance.Choice of reaction buffers, with or without MgCl2.Non-proofreading Polymerase
Catalog No. NC2160335
Encompass
EconoTaq DNA Polymerase is provided with a choice of 10X Reaction Buffer with Mg++ (with Mg++); or 10X Reaction Buffer without Mg++ and a separate tube of 25 mM MgCl2 (separate Mg++).
Catalog No. NC2160335 Supplier LGC Genomics Llc Supplier No. 300321*
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
EconoTaq DNA Polymerase is provided with a choice of 10X Reaction Buffer with Mg++ (with Mg++); or 10X Reaction Buffer without Mg++ and a separate tube of 25 mM MgCl2 (separate Mg++).