missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Economy Vinyl Floor Tape 3 in. W x 108 ft. L PVC
Description
- Affordable vinyl floor tape for everyday use.
- Easy to apply.
- Suitable for facility marking and Lean/5S programs.
- Meets OSHA and ANSI guidelines for marking hazards and aisleways.
- Industrial-grade vinyl for light-to-medium traffic areas.
- Rubber-based adhesive adheres to most clean, dry surfaces.
Spécifications
Spécifications
| Couleur | Yellow, Black |
| Type d’adhésif | Rubber |
| Matériau | PVC |
| Type de produit | Floor Marking Tape |