Learn More
Eisco™ Economy Spectroscope Tubes
Catalog No. S24884
Change view
Click to view available options
Quantity:
24 Pk.
6 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24884 24 Pk.
S24885 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24884 Supplier Eisco™ Supplier No. PH0596CBT24
Only null left
Economy Spectroscope Tubes for identifying constituent wavelengths of visible light in many sources.
  • Allows user to view through slit spectrum of light decomposed by optical system.
  • Available as pack of 6 or 24.
Specifications
Product Type Spectroscope Tube
For Use With (Application) Identifying constituent wavelengths of visible light in many sources
Quantity 24 Pk.