Learn More
Eisco™ Economy Spectroscope Tube
Catalog No. S14007
Catalog No. S14007 Supplier Eisco™ Supplier No. PH0596CBTPK15
Designed to let the user look through a slit spectrum of light decomposed by an optical system. EISCO Economy Spectroscope Tubes allows students to quickly identify constituent wavelengths of visible light in many sources,
  • Basic spectroscopes able to view variable light spectra
  • Resulting spectra as viewed through the grating introduces students to many properties of visible light and optics
  • Hold it up to different lights to observe which wavelengths are present
  • Light enters through the slit and is split by the diffraction grating at the other end
  • Perfect activity for students at school or kids at home An adjustable thin slit cap rotates to produce diffraction patterns in the desired direction
Specifications
Product Type Spectroscope Tube
For Use With (Application) Identify constituent wavelengths of visible light in many sources.
Includes Adjustable thin slit cap