missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Economy Aluminum Tuning Fork
Economy aluminum tuning forks are designed for sound wave lessons and exploration
Supplier: Eisco™ PH0737C
Description
- Aluminum tuning fork
- Economy priced
- Frequency within ± 5%
Chemical Identifiers
| Tuning Fork | |
| Aluminum | |
| Sound Wave Lessons and Exploration |
Specifications
| Tuning Fork | |
| Aluminum | |
| Sound Wave Lessons and Exploration | |
| 320 Hz |