Learn More
Eisco™ Economy 7 Piece Dissecting Set
Catalog No. S14004
Change view
Click to view available options
Size:
7 pieces
Catalog No. Size
S14004 7 pieces
1 options
Catalog No. S14004 Supplier Eisco™ Supplier No. BI0152ECO
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless steel dissection set, consisting of 7 instruments for accurate dissection.
Supplied in leather storage case:
  • (1) Scissors
  • (1) Forceps
  • (1) Straight needle with wooden handle
  • (1) Bent needle with wooden handle
  • (1) Glass pipet with rubber dropper
  • (1) Scalpel handle with sterile blade
  • (1) 6 in. plastic ruler
Specifications
Product Type Dissection Set
Disposable Reusable
For Use With (Application) Basic dissection practice
Includes 1 pointed tip scissor (4 in.), 1 blunt end forceps (5 in.), 1 straight needle with 4 in. wooden handle, 1 curved needle with 4 in. wooden handle, 1 scalpel handle #4, 1 scalpel blade #20, 1 glass dropper (4 in.) with rubber teat, 1 acrylic 6 in. ruler
Size 7 pieces
Weight (English) 0.5 lb.
Sterility Non-sterile