missing translation for 'onlineSavingsMsg'
Learn More

Economical pH Test Strips

Numéro de catalogue. S6115
Change view
Click to view available options
pH Range:
0 to 1.5
0 to 2.5
11.5 to 14
3 to 6
5 to 9
7 to 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. pH Range
S6115 7 to 10
S6112 0 to 2.5
S6117 11.5 to 14
S6111 0 to 1.5
S6113 3 to 6
S6114 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. S6115 Fournisseur Customer Channels Group (Ccg) Code fournisseur PH70101B50
Il en reste null

Line of pH test strips incorporates the most up-to-date technology.

  • Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
  • Stabilized to provide the most accurate color determinations
  • Color charts printed on each pack for quick and easy identification
  • Each pack includes 50 strips

Spécifications

pH Range 7 to 10
For Use With (Application) Stabilized to provide user with most accurate color determination
Graduations 0.5
Quantity 50 Bag
Type pH Test Strip