Learn More
United Scientific™ EcoLift Lab Jack
Description
The EcoLift™ Lab Jacks are constructed of powder coated stainless steel. The height can be easily adjusted with a simple turn of the comfortable molded knob. The jacks are available in three sizes to fit various laboratory applications.
- Powder coating provides corrosion resistance
- Stable and durable
- Height is easily adjustable
Specifications
Specifications
| Material | Powder Coated Stainless Steel |
| Height (Metric) | 6.35 to 15 cm |
| Product Type | Lab Jack |
Cancer and Reproductive Harm