missing translation for 'onlineSavingsMsg'
Learn More

ZING™ Enterprises Eco Fire Blanket - Tote Set

Catalog No. 18002982
Encompass
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18002982 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18002982 Supplier ZING™ Enterprises Supplier No. EPD7230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Over sized blanket is fire resistant and used to smother flames and cover injured victims.

ZING Eco Fire Blanket and Hanging Tote is made from 100% recycled materials. Over sized blanket is fire resistant and used to smother flames and cover injured victims. Blanket and Tote with hanging grommets is ideal for use in First Aid Stations, Disaster Kits, Plants, Offices, Schools, Fire Trucks, Police Cars, Utility Trucks, Ambulances and Rescue Vehicles. Sold as a set. The blanket size is 96 in.x60 in. and the complete kit size of 23 in.Hx23 in.Wx4 in.D

Specifications

Product Type Fire Blanket
Color Red
Material Recycled Materials
Dimensions (L x W) 96 x 60 in. (23 x 4 in. in carrier bag)
For Use With (Application) Safety
Includes 1 Blanket and Holder with Corner Holes Mounting
Length (English) 23 in.
Width (English) 4 in.