missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Eco Best Technology™ Disposable Nitrile Gloves

Catalog No. 296367049
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Catalog No. Size
296367049 X-Small
296367050 Small
296367051 Medium
296367052 Large
296367053 X-Large
296367054 2X-Large
6 options

Catalog No. 296367049

Supplier: SHOWA™ 6112PFXS

Only null left

Eco Best Technology™ (EBT) biodegradable nitrile disposable gloves provide comfort, dexterity and performance across a wide range of industrial applications.

  • Same performance as non-EBT nitrile disposable gloves
  • Eco Best Technology™ (EBT) achieved 82% biodegradation in 386 days
  • Decreased risk of allergies
  • Second-skin fit and feel
  • Recommended for industrial applications including food, pharmaceutical, utilities, construction, agricultural and automotive

Compliance

Data Testing is available upon request.

Specifications

Certifications/Compliance ASTM D5511-18, ASTM D5526
Certifications/Compliance (Europe) EN 374-2:2003, EN 374-1:2016/Type B, EN 374-5:2016
Applications Industrial
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Size X-Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6112PFXS
Show More Show Less