Learn More
Torrey Pines Scientific™ EchoTherm™ Orbital Mixing Chilling/Heating Dry Bath
Used for any molecular biology lab mixes samples as they are heated or chilled with a variable speed orbital mixer
Supplier: Torrey Pines Scientific™ SC20
Description
EchoTherm™ SC20 Orbital Mixing Chilling/Heating Dry Bath is compact, versatile, reliable and easy to use. The built-in orbital mixer is designed with friction free hardened, blued, tempered steel leaf spring bearings that support the plate and sample blocks. Units come complete with instruction manual, universal bench top power supply for use anywhere in the world and AC line cord for the country of use. Sample blocks and covers are available as accessories.
- Orbital mixing with speeds from 200 to 1000rpm
- Energy-efficient Peltier design
- Digital control to 1°C from -10° to 100°C
- Control at room temperature
- Thirty-day count down timer with audible alarm and user settable Auto-Off
- RS-232 I/O port for control or data collection
- Built-in data logger
- Electronic calibration
- Small compact size uses very little bench space
- 12V DC operation. Universal power supply provided
- Range: -10° to 100°C
- Readability: 1°C
- Accuracy:±1°C
- Stability:±1°C
- Sensor: Platinum RTD
- Controller Type: PID
- Uniformity:±0.5°C
- Mixing samples in centrifuge tubes, assay plates, PCR plates, vials, test tubes or other containers while heating or chilling
- Maintaining 14°C for ligation reaction
- Maintaining 17°C for storing oocytes
- Storing samples at ice bucket temperatures without the mess
- Incubating samples at 37°C
- Storing enzyme or DNA libraries at your workstation
- Enzyme reactions and deactivations
Compliance
UL, CSA, and CE compliant
Specifications
| UL, CSA, CE | |
| Non-programmable | |
| ±1°C | |
| 19.69 cm | |
| 10.8 cm | |
| 36.83 cm | |
| 10.9 kg | |
| 100/265 V |
| -10°C to +100°C | |
| 200 to 1000rpm | |
| 7.75 in. | |
| 4.25 in. | |
| 14.5 in. | |
| 24 lb. | |
| 100/265 V 50/60 Hz | |
| 50/60 Hz |