Learn More
Beckman Coulter EchoQualified 384-well Cyclic Olefin Copolymer (COC) Source Microplate, Low Dead Volume, Clear, Non-sterile
Catalog No. NC1731983
Encompass
EchoQualified 384-Well Low Dead Volume (LDV) Source Microplates are designed to minimize dead volume when using the EchoLiquid Handler. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performanceand DMSO storage. Each lot of microplates is tested and certified to meet stringent performance specifications. The 384-Well Low Dead Volume (LDV) plate is designed to minimize dead volume when using the Echoliquid handling system. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performance and DMSO storage. Each lot of microplates is tested and certified to meet stringent performance specifications.
Catalog No. NC1731983 Supplier Beckman Coulter Supplier No. 00112782
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
EchoQualified 384-Well Low Dead Volume (LDV) Source Microplates are designed to minimize dead volume when using the EchoLiquid Handler. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performanceand DMSO storage. Each lot of microplates is tested and certified to meet stringent performance specifications. The 384-Well Low Dead Volume (LDV) plate is designed to minimize dead volume when using the Echoliquid handling system. This microplate is constructed of Cyclic Olefin Copolymer (COC) for optimal acoustic transfer performance and DMSO storage. Each lot of microplates is tested and certified to meet stringent performance specifications.