Learn More
MilliporeSigma™ ech2o™ Oxidation Lamp
Catalog No. ZIQUVLPA1
Change view
Click to view available options
For Use With (Equipment):
Milli-Q™ IQ 7000, Milli-Q™ IQ 7003/05/10/15 Systems
Catalog No. For Use With (Equipment)
ZIQUVLPA1 Milli-Q™ IQ 7000, Milli-Q™ IQ 7003/05/10/15 Systems
1 options
Catalog No. ZIQUVLPA1 Supplier MilliporeSigma™ Supplier No. ZIQUVLPA1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement mercury-free UV lamp for photooxidation of organic compounds
  • Used with Milli-Q™ IQ 7000 and Milli-Q™ IQ 7003/05/10/15 systems
Specifications
Product Type Oxidation Lamp
For Use With (Equipment) Milli-Q™ IQ 7000, Milli-Q™ IQ 7003/05/10/15 Systems
For research purposes only