missing translation for 'onlineSavingsMsg'
Learn More

Ecdysterone

Catalog No. aaj63091mf
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63091MF 50 mg
AAJ63091MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63091MF Supplier Thermo Scientific Chemicals Supplier No. J63091MF
Only null left

An insect steroid hormone that induces apoptosis | CAS: 5289-74-7 | C27H44O7 | 480.64 g/mol

Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

Solubility
Soluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 5289-74-7
Molecular Formula C27H44O7
Molecular Weight (g/mol) 480.64
MDL Number MFCD00036740
InChI Key NKDFYOWSKOHCCO-YPVLXUMRSA-N
Synonym 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson
PubChem CID 5459840
ChEBI CHEBI:16587
IUPAC Name (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one
SMILES CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Melting Point 242°C to 244°C
Color White
Quantity 50 mg
Beilstein 1917578
Merck Index 14,3491
Solubility Information Soluble in alcohol
Formula Weight 480.64
Physical Form Powder
Chemical Name or Material Ecdysterone
RTECSNumber FZ8060000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only