missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human Treg Phenotyping Panel & Staining Buffers

Catalog No. A42924
Encompass
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42924 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42924 Supplier Invitrogen™ Supplier No. A42924

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen™ eBioscience™ Essential Human Treg Phenotyping Panel and Staining Buffers are designed for flow cytometry-based identity testing of regulatory T-cells (Tregs).

Phenotyping Panel and Staining Buffers leverages high quality eBioscience™ antibodies and fixation/permeabilization reagents (for the intracellular marker FOXP3) and includes a ready-to-use protocol for experimental set-up, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies and supporting reagents designed for flow cytometry
  • Highly referenced antibodies―includes one of the most referenced clones for FOXP3
  • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization

Specifications

Content And Storage
  • CD4, CD25, CD127, and FOXP3 antibodies, store at 2–8°C
  • FOXP2/TF Staining Buffer set, store at 2–8°C
Format Kit
Quantity 100 reactions
Detection Method Fluorescence
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line eBioscience
Product Type Phenotyping Panel and Staining Buffer

For Research Use Only. Not for use in diagnostic procedures.