missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human T-Cell Phenotyping Panel

Catalog No. A42806
Encompass
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42806 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42806 Supplier Invitrogen™ Supplier No. A42806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen™ eBioscience™ Essential Human T-Cell Phenotyping Panel is designed for flow cytometry-based identity testing of T-cells

Essential Human T-Cell Phenotyping Panel everages high quality eBioscience™ antibodies and a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies designed for flow cytometry
  • Highly referenced antibodies―eBioscience™ portfolio contains trusted fluorochrome-conjugated antibodies for immunology and oncology research
  • Turnkey kit and protocol―contains standardized gating recommendations needed to perform flow cytometry characterization

Specifications

Content And Storage CD3, CD4, CD8, CD62L, and CCR7 antibodies, store at 2°C to 8°C
Format Kit
Quantity 100 reactions
Detection Method Fluorescence
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line eBioscience
Product Type Phenotyping Panel

For Research Use Only. Not for use in diagnostic procedures.