missing translation for 'onlineSavingsMsg'
Learn More

Eberbach Container for Waring™ Blenders

Catalog No. 1450925
Encompass
Change view
Click to view available options
For Use With (Equipment):
Waring Lab Blender
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
1450925 Waring Lab Blender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1450925 Supplier Eberbach Supplier No. E8580.00

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Semi-micro container

  • Stainless steel container
  • Press-fit stainless steel cover
  • Total Capacity: 360mL
  • Working Capacity: 25mL min.; 250 mL max.

Specifications

Product Type Container
Capacity (Metric) 360 mL
Material Stainless Steel
For Use With (Equipment) Waring Lab Blender
Includes Stainless Steel cover