missing translation for 'onlineSavingsMsg'
Learn More

Eberbach Blender Container

Catalog No. 1450924
Encompass
Change view
Click to view available options
For Use With (Equipment):
Waring™ blender
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
1450924 Waring™ blender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1450924 Supplier Eberbach Supplier No. EBB E8470.00

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Type Container
Capacity (Metric) 500 mL
Size Small
Diameter (English) 3.75 in.
Diameter (Metric) 9.52 cm
Material Glass
For Use With (Equipment) Waring™ blender
Includes Stainless Steel blades, metal screw cover