missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Easystrip™ Plus Tube Strip with Attached Ultra Clear Caps

Catalog No. AB2005SMP
Change view
Click to view available options
Color:
Optical
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
AB2005SMP Optical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2005SMP Supplier Thermo Scientific™ Supplier No. AB2005SMP

Specifications

Certifications/Compliance Certified human DNA, RNase and DNase free
Color Optical
Storage Requirements Room Temperature
Quantity 5 Strips
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Closure Type Flat Cap
Product Type 8-strip Tube