missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One EASYseal™ Adhesive Microplate Sealer
Description
- Coated with acrylate adhesive
- Coverage/storage of microplates (e.g in ELISA applications)
- Protection against evaporation and contamination
- Suitable for optical measurements
Specifications
Specifications
| Color | Transparent |
| Sterility | Non-sterile |
| No. per Case | 100 |
| For Use With (Application) | ELISA |
| Product Type | Sealer |
| Adhesive | Acrylate |
| Plate Compatibility | Microplate |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.