Learn More
StatLab EasyCut Decal Solution
Catalog No. DCECDGAL
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
DCECDGAL 1 gal.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. DCECDGAL Supplier StatLab Supplier No. DCECDGAL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or if inhaled
StatLab EasyCut Decal Solution is a medium strength decalcifier that can be used for all routine decalcification needs
  • It is formulated to provide less aggressive decalcification than rapid decalcifying solutions and complete demineralization in a timely manner
  • EasyCut Decal Solution offers an excellent balance between decalcification time versus protection of non-calcium containing tissue elements
Specifications
Product Type Decalcifying Solution
Quantity 1 gal.
Solution Type Decalcifier
Form Solution