missing translation for 'onlineSavingsMsg'
Learn More

Contec™ EasyCurve™ Mop Heads

Catalog No. p-4909793
Change view
Click to view available options
Material:
Knitted Polyester Microfiber, Hydrophilic Urethane Foam
Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Product Type:
Mop Head
Sealed Edge Mop Head
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Sterility
19159242 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Mop Head Non-sterile
19159243 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Mop Head Sterile
19138940 Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge Mop Head Non-sterile
19456891 Knitted Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge Mop Head Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19159242 Supplier Contec™ Supplier No. EZMP0300
Only null left

Lightweight mop for ceilings, walls, and floors. Choose from non-sterile or validated sterile versions. Mop heads are autoclavable. The EasyCurve™ Mop Heads use curved mop faces with pivoting connectors for easier surface contact at a variety of angles.

Contec's EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users smooth and easy mopping for better particle pick up and contamination removal. The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability, making the EasyCurve an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve Mop are autoclavable.

Features & Benefits

  • All components are autoclavable which is ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal

Specifications

Color Code White
Product Type Mop Head
Sterility Non-sterile
Autoclavable Autoclavable
Size 36 x 11 x 2 cm (14 x 4.25 x 0.75 in.)
Disposable Single-use
Quantity 4/Bag, 12 Bags/Cs.
For Use With (Equipment) EasyCurve Mop System
For Use With (Application) Designed for use in pharmaceutical and medical device manufacturing plants, hospitals, compounding pharmacies and other critical or controlled environments
Color-coded No
Material Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Show More Show Less