Learn More
Research Products Corporation Easy Dip Staining Jar, White, 6 per Package
Catalog No. 501977961
Encompass
Unique slide staining system features a square staining jar and a vertical slide rack. Both components are made from heavy duty acetal polymer resistant to alcohol and Xylene. Staining jar has a wide stable base and a recessed interior cavity with a reduced volume capacity of 80ml. Slide staining rack holds up to 12 slides with dimensions of 25 x 75mm, 25x76mm or 1" x 3". Slides are held vertically for easy insertion and removal. Rack is designed for quick drying of slides in a microwave oven or at temperatures up to 120C. Jars available in five different colors for sample identification.
Jars available in five different colors for sample identification. Each jar is supplied with clip for quick and easy connection of staining jars.
Slide staining rack is sold separately.
Catalog No. 501977961 Supplier Research Products Corporation Supplier No. 247835W
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Unique slide staining system features a square staining jar and a vertical slide rack. Both components are made from heavy duty acetal polymer resistant to alcohol and Xylene. Staining jar has a wide stable base and a recessed interior cavity with a reduced volume capacity of 80ml. Slide staining rack holds up to 12 slides with dimensions of 25 x 75mm, 25x76mm or 1" x 3". Slides are held vertically for easy insertion and removal. Rack is designed for quick drying of slides in a microwave oven or at temperatures up to 120C. Jars available in five different colors for sample identification.
Jars available in five different colors for sample identification. Each jar is supplied with clip for quick and easy connection of staining jars.
Slide staining rack is sold separately.