missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Earle's Balanced Salts Solution, without phenol red

Catalog No. p-7057918
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67559AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67559AP Supplier Thermo Scientific™ Supplier No. J67559AP
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Synonym EBSS