Learn More
World Precision Ear Punch 1mm diameter
Supplier: World Precision WP500076
Ring Handled Ear Punches are commonly used in a laboratory setting for tagging rodents for easy identification. Ear punches can create a distinctive hole or pattern in the ear of a rodent. Ear punches are relatively non-invasive compared with ear tags, tattoos and other methods. They create a hole in the ear without causing significant harm or discomfort to the animal. Ear punches permanent mark your rodents, without ever fading or require further maintenance. You can easily identify individual animals throughout their lifespan. The distinctive pattern of the ear punch can be easily seen and identified by laboratory personnel. Ear punches are relatively inexpensive compared to other tagging methods. Ring Handled Ear Punches offer several advantages over simple ear punches. The ring handles give you greater control over the punching action, which means you get more consistent results and fewer misalignments. .