missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Single-Drum Spill Containment Unit
GREENER_CHOICE

Catalog No. 189994473
Change view
Click to view available options
Capacity (English):
65 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
189994473 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189994473 Supplier Eagle™ Supplier No. 1612

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for forklift use

Ideal for use as spill containment unit, mobile pumping station, or waste collection station.

  • Provides secondary spill containment of hazardous materials when used with drums up to 55 gal. (208L)
  • Constructed of high-density polyethylene with UV inhibitors for superior strength and excellent chemical resistance
  • Nestable for easy storage

Order Info

Compatible with 18-869 drum dolly (sold separately). Capacity: 65 gal. (246L). Weight: 30 lb. (13.6kg). Dimensions: 31W x 33 in. H (78.7 x 84cm).

Specifications

Certifications/Compliance EPA, 40 CFR 264.175
Color Yellow
Capacity (English) 65 gal.
Capacity (Metric) 246 L
Material HDPE
Type Single Drum Containment Unit
For Use With (Application) Forklift use
For Use With (Equipment) For use as spill containment unit, mobile pumping station, or waste collection station
Holds 1 Drum
Includes 0.75 in. drain plug, black grating with 1.75 in. openings, 78.7 x 83.8cm
Resistant Chemical
Length (English) 31 in.
Length (Metric) 78.7 cm
Height (English) 33 in.
Height (Metric) 83.8 cm
Weight (English) 30 lb.
Weight (Metric) 13.6 kg
Show More Show Less