missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Polyethylene Oily Waste Cans
GREENER_CHOICE

Designed for temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion

Supplier:  Eagle™ 935FLY

 View more versions of this product

Catalog No. 19038453


Only null left

Description

Description

Durable, fire-safe cans won't rust or dent and resist most chemical compounds.

  • Cans feature foot treadle that opens cover; self-closing lid
  • Choice of red or high-visibility yellow

Certifications

FM approved.

Compliance

Meet OSHA 29CFR1910.108 and 20CFR1910.106 requirements.

Specifications

Chemical Identifiers

FM, OSHA 29 CFR 1910.108, 20 CFR 1910.106
1 Ea.
For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Oily Waste Can
Self Closing
Foot lever
HDPE

Specifications

38 L
18 in.
45.7 cm
Yellow
1 Ea.
6.3 kg
Foot lever
22 in.
HDPE
FM, OSHA 29 CFR 1910.108, 20 CFR 1910.106
Self Closing
10 gal.
46 cm
14 lb.
55.8 cm
For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
18 in.
Oily Waste Can
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions