Learn More
Eagle™ Drum Covers
Catalog No. 18885B
Change view
Click to view available options
For Use With (Equipment):
Closed and Open-head Drums
Open-head Drums
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18885B Open-head Drums
18885A Closed and Open-head Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18885B Supplier Eagle™ Supplier No. 1667
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Made of high-density polyethylene that resists most chemicals, acids and corrosives. Covers have snap-on design for positive closure. Snap on or off without cracking or splitting. Dome shape protects drum from water or other contaminants. Available for closed head and open head drums.
Order Info
Fit 55 gal. (208L) metal drums.
Specifications
For Use With (Equipment) Open-head Drums